Změnit zobrazení
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
12 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 5 710,00 Kč |
O této položce
NACRES:
NB.24
UNSPSC Code:
41123100
5 710,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimaterial
blue
packaging
pack of 12 ea (dry with glycerol)
manufacturer/tradename
Spectrum Labs G235071
pore size
100 kDa MWCO
capacity
10 mL
General description
- 95 - 98% Sample Recovery
- Highest Membrane Purity
- Superior Handling & Leak Prevention
- Volume Specific Dilution Control
- Complete & Ready-to-Use Packaging
Spectrum′s proprietary Biotech Grade CE is a low protein-binding synthetic membrane available in 4 precise MWCO′s (color coded for MWCO variants) with no heavy metal and sulfide contaminants. The cylindrical tubing geometry prevents sample dilution (associated with cassette-type devices) and provides open access for total volume retrieval. Only the Float-A-Lyzer® G2 assures a 95-98% sample recovery while maintaining 99% sample purity and <1% sample dilution.
The leak-proof screw-on cap with sealing o-ring provides easy access with included pipette for loading, in-process testing and sample retrieval, without the risk of needle punctures. The included floatation ring improves sample buoyancy and vertical orientation during dialysis. The sleek design allows multiple samples to be dialyzed in the same reservoir.
No additional kits, buoys or accessories are required. The Float-A-Lyzer G2 is packaged dry and ready-to-go with all components included. Simply rinse with water, load the sample with a pipette, and dialyze.
Spectrum Laboratories, Inc. introduces the Spectra/Por® Float-A-Lyzer® G2, the next generation of Ready-to-Use laboratory dialysis devices featuring a Ultra-pure Biotech Cellulose Ester (CE) Membrane
Legal Information
Float-A-Lyzer is a registered trademark of Spectrum Laboratories, Inc.
Spectra/Por is a registered trademark of Spectrum Laboratories, Inc.
1 of 1
Tato položka | |||
|---|---|---|---|
| description blue, 10 mL, MWCO 100 kDa | description blue, 1 mL, MWCO 100 kDa | description blue, 5 mL, MWCO 100 kDa | description yellow, 5 mL, MWCO 8-10 kDa |
| packaging pack of 12 ea (dry with glycerol) | packaging pack of 12 ea (dry with glycerol) | packaging pack of 12 ea (dry with glycerol) | packaging pack of 12 ea (dry with glycerol) |
| pore size 100 kDa MWCO | pore size 100 kDa MWCO | pore size 100 kDa MWCO | pore size 8-10 kDa MWCO |
| manufacturer/tradename Spectrum Labs G235071 | manufacturer/tradename Spectrum Labs G235035 | manufacturer/tradename Spectrum Labs G235059 | manufacturer/tradename Spectrum Labs G235055 |
| capacity 10 mL | capacity 1 mL | capacity 5 mL | capacity 5 mL |
| material blue | material blue | material blue | material yellow |
Still not finding the right product?
Explore all of our products under Spectra-Por® Float-A-Lyzer® G2
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?