Změnit zobrazení
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
1 pkg | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 1 570,00 Kč |
O této položce
NACRES:
NB.01
UNSPSC Code:
41121500
Sterility:
non-sterile
1 570,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlisterility
non-sterile
packaging
pkg of 10 ea
manufacturer/tradename
BRAND 26535
General description
The accu-jet®S / pro / macro pipette controller is designed to safeguard your equipment with its robust PP housing and integrated interior membrane that effectively blocks liquid. This protective feature ensures the longevity of your pipette controller while maintaining a clean and safe working environment.
Other Notes
BR26535 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
accu-jet is a registered trademark of BRAND GMBH + CO KG
1 of 1
Tato položka | |||
|---|---|---|---|
| description spare membrane filter 0.2 μm, non-sterile | description polyethersulfone membrane, sterile, pore size 0.22 μm, membrane diam. 70 mm, funnel capacity 1000 mL, case of 12 | description spare membrane filters 0.2 μm, sterile | description cellulose acetate membrane, sterile, pore size 0.22 μm, membrane diam. 70 mm, funnel capacity 1000 mL, case of 12 |
| sterility non-sterile | sterility sterile | sterility sterile | sterility sterile |
| manufacturer/tradename BRAND 26535 | manufacturer/tradename Corning 431174 | manufacturer/tradename BRAND 26530 | manufacturer/tradename Corning 430015 |
| packaging pkg of 10 ea | packaging case of 12 | packaging pkg of 1 ea | packaging case of 12 |
Still not finding the right product?
Explore all of our products under BRAND® accu-jet® pro pipette controller membrane filters
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?