Přeskočit na
Z326704
BRAND® BLAUBRAND® ETERNA volumetric flask
volume 5 mL, accuracy: 0.04 mL
Synonyma:
analytical flask, calibration flask with stopper, glass flask, measuring flask, volumetric flask
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
Vám/Skladová položkaDostupnostCena
2 ea
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
1 100,00 Kč
O této položce
NACRES:
NB.43
UNSPSC Code:
41121804
Capacity:
5 mL
Material:
borosilicate glass , wide-neck
Packaging:
pack of 2
1 100,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
material
borosilicate glass , wide-neck
joint
joint ST/NS neck10/19, joint ST/NS10/19
description
calibrated to contain (TC, EX)
sterility
non-sterile
feature
accuracy class A (DIN EN ISO 1042, calibrated to contain (TC, EX)), closure type PP Stopper
packaging
pack of 2
manufacturer/tradename
BRAND
volume measuring range
5 mL, accuracy: 0.04 mL
capacity
5 mL
General description
Class A, conformity certified. Made from Boro 3.3, Supplied with a square grip PP stopper. Marks and inscriptions in ETERNA amber diffusion stain, particularly resistant to aggressive cleaning methods. TO DIN EN ISO 1042.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
Other Notes
BR36838 (India)
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Porovnat podobné položky
Zobrazit úplné porovnání
Zobrazit rozdíly
1 of 1
Tato položka
Z326801Z326755Z326747
description
volume 5 mL, accuracy: 0.04 mL
description
volume 500 mL, accuracy: 0.25 mL
description
volume 50 mL, accuracy: 0.08 mL, neck joint: ST/NS 14/23
description
volume 50 mL, accuracy: 0.06 mL, neck joint: ST/NS 12/21
feature
accuracy class A (DIN EN ISO 1042, calibrated to contain (TC, EX)), closure type PP Stopper
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In)., closure type PP Stopper
feature
accuracy class A (DIN EN ISO 1042, calibrated to contain (TC, EX)), closure type PP Stopper
feature
accuracy class A (DIN EN ISO 1042, calibrated to contain (TC, EX)), closure type PP Stopper
capacity
5 mL
capacity
500 mL
capacity
50 mL
capacity
50 mL
manufacturer/tradename
BRAND
manufacturer/tradename
BRAND
manufacturer/tradename
BRAND
manufacturer/tradename
BRAND
material
borosilicate glass , wide-neck
material
borosilicate glass
material
borosilicate glass , wide-neck
material
borosilicate glass
joint
joint ST/NS10/19, joint ST/NS neck10/19
joint
joint ST/NS neck19/26
joint
joint: ST/NS neck14/23
joint
joint: ST/NS neck12/21
packaging
pack of 2
packaging
pack of 2
packaging
pack of 2
packaging
pack of 2
Still not finding the right product?
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)