Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 728,00 Kč |
O této položce
728,00 Kč
material
nickel closure, white
feature
Holds 100 x slides 76x26 mm
manufacturer/tradename
Heathrow Scientific HS15994R
L × W × H
208 × 175 × 34 mm
General description
Features and Benefits
- molded, rigid boxes securely hold slides in grooved slots
- durable ABS plastic boxes are available wth either cork or foam lining
- both the lining and the slots are numbered to correspond to the slide inventory sheet
- closes securely with a rust-resistant, nickel-plated clasp and pin hinges
1 of 1
Tato položka | |||
|---|---|---|---|
| description 100 Place, foam lining, white | description 100 Place, cork lining, green | description 100 Place, cork lining, blue | description 100 Place, cork lining, black |
| feature Holds 100 x slides 76x26 mm | feature Holds 100 x slides 76x26 mm | feature Holds 100 x slides 76x26 mm | feature Holds 100 x slides 76x26 mm |
| manufacturer/tradename Heathrow Scientific HS15994R | manufacturer/tradename Heathrow Scientific HS15994B | manufacturer/tradename Heathrow Scientific HS15994A | manufacturer/tradename Heathrow Scientific HS15994G |
| material nickel closure, white | material green , nickel closure | material blue , nickel closure | material black , nickel closure |
| L × W × H 208 × 175 × 34 mm | L × W × H 208 × 175 × 34 mm | L × W × H 208 × 175 × 34 mm | L × W × H 208 × 175 × 34 mm |
Still not finding the right product?
Explore all of our products under Premium Slide box
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?