Změnit zobrazení
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 963,00 Kč |
O této položce
NACRES:
NB.35
UNSPSC Code:
41121812
Manufacturer/tradename:
Heathrow Scientific HS14521B
963,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimanufacturer/tradename
Heathrow Scientific HS14521B
D × H
57 mm × 12 mm , meduim
General description
designed for general-purpose weighing, dispensing, storage, food processing, evaporating, or drying.
Boats also make good dust covers for open vessels.
Sturdy yet lightweight boats feature a smooth, flat bottom for sample handling and a flanged, crimped wall for rigidity. Finger tab handle can be etched for identification prior to weighing.
3 sizes
Packs of 100
Boats also make good dust covers for open vessels.
- All weigh boats are FDA/food-grade compliant
- Disposable
- Aluminum, Weighing Boats
Sturdy yet lightweight boats feature a smooth, flat bottom for sample handling and a flanged, crimped wall for rigidity. Finger tab handle can be etched for identification prior to weighing.
3 sizes
Packs of 100
1 of 1
Tato položka | |||
|---|---|---|---|
| manufacturer/tradename Heathrow Scientific HS14521B | manufacturer/tradename Heathrow Scientific HS14521C | manufacturer/tradename Heathrow Scientific HS14521A | manufacturer/tradename Sigma-Aldrich |
| D × H 57 mm × 12 mm , meduim | D × H 70 mm × 12 mm , large | D × H 43 mm × 12 mm , small | D × H - |
Still not finding the right product?
Explore all of our products under Aluminium weighing boat
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?