Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 kit | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 4 510,00 Kč |
O této položce
4 510,00 Kč
description
Tricorn glass columns may be used with aqueous buffer solutions and detergents at pH conditions between 1 and 14, and with nearly all organic solutions commonly used in chromatography.
feature
Ethylene propylene diene (EPDM) Polyethylene (PE) , Kit Contents: 5 x Top filter 5 x Bottom filter 5 x O-ring 7 x 1 mm (EPDM) , Tricorn 5 Columns, Wetted Material: Ethylene propylene diene (EPDM) Polyethylene (PE)
manufacturer/tradename
Cytiva 29053586
parameter
4 °C min. temp., 60 °C max. temp.
General description
Tricorn columns incorporate modern design and high performance to increase the user friendliness and reliability of the chromatography column. Tricorn columns are designed for high resolution liquid chromatography of biomolecules performed at medium to high pressures.
Features and Benefits
- High performance glass columns designed for reliable high resolution, reproducibility and durability
- New longer adaptor for increased adjustment of bed height
- Modern, integrated design – simple to assemble, pack, operate, and maintain
- Broad range of column dimensions – suitable for many different purification needs
- Safety first – glass tubes coated with plastic film to protect laboratory personnel if the column is dropped or operated outside its specifications
- Locking ring – prevents accidental compression of media bed
- Separate packing connector – allows simple column packing for a broad range of media
1 of 1
Tato položka | |||
|---|---|---|---|
| description Tricorn 5 Columns | description Tricorn 10 Columns | description joint: 10/100 | description Tricorn 10 Column |
| manufacturer/tradename Cytiva 29053586 | manufacturer/tradename Cytiva 29053612 | manufacturer/tradename Cytiva 18-1153-15 | manufacturer/tradename Cytiva 18-1153-10 |
| feature Wetted Material: Ethylene propylene diene (EPDM) Polyethylene (PE) , Tricorn 5 Columns, Kit Contents: 5 x Top filter 5 x Bottom filter 5 x O-ring 7 x 1 mm (EPDM) | feature Tricorn 10 Columns, Wetted Material: Ethylene propylene diene (EPDM) Polyethylene (PE) , Kit Contents: 5 x Top filter 5 x Bottom filter 5 x O-ring 7 x 1 mm (EPDM) | feature - | feature Tricorn 10 Column |
| parameter 4 °C min. temp., 60 °C max. temp. | parameter 4 °C min. temp., 60 °C max. temp. | parameter - | parameter - |
| description Tricorn glass columns may be used with aqueous buffer solutions and detergents at pH conditions between 1 and 14, and with nearly all organic solutions commonly used in chromatography. | description Tricorn glass columns may be used with aqueous buffer solutions and detergents at pH conditions between 1 and 14, and with nearly all organic solutions commonly used in chromatography. | description - | description - |
Tricorn filter kit for MabSelect PrismA | - | Tricorn glass tube for medium to high pressure applications, high performance Tricorn glass tube for biomolecules, medium pressure Tricorn glass column, Tricorn glass tube chromatography column, integrated design Tricorn glass column for research, Tricorn 10/100 glass column design and performance, Tricorn 10/100 glass tube for high resolution chromatography, Tricorn glass chromatography column 10/100, high resolution Tricorn glass chromatography tube, Tricorn glass tube with adjustable bed height, Tricorn glass columns for lab use, reproducible chromatography with Tricorn glass tube, Tricorn 10/100 chromatography tube assembly instructions, durable Tricorn glass tube for liquid chromatography | durable Tricorn 10 column bottom unit, Tricorn 10 Bottom Unit high performance, Tricorn Bottom Unit lab-grade chromatography, Tricorn column bottom unit for liquid chromatography, Tricorn Bottom Unit chromatography accessories, Tricorn Bottom Unit chromatography column, Tricorn Bottom Unit assembly and operation, Tricorn Bottom Unit integrated design features, Tricorn Bottom Unit for biomolecule separation, reliable Tricorn Bottom Unit for research, Tricorn column bottom unit for medium to high pressures, high resolution Tricorn liquid chromatography column, medium pressure Tricorn column adapter |
Still not finding the right product?
Explore all of our products under Tricorn Filter Kit
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?