Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
100 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 1 260,00 Kč |
O této položce
1 260,00 Kč
material
colorless , polycarbonate
sterility
non-sterile
packaging
pkg of 100 ea
manufacturer/tradename
Grace Bio-Labs 714022
L × W × thickness
22 mm × 40 mm × 0.25 mm
General description
HybriSlips are flexible, thin (0.25 mm) covers that minimize fluid friction and facilitate uniform reagent distribution. RNase free, hydrophobic covers will not trap or bind probes to their surfaces like coverglass. Product is ready-to-use without pretreatment. Working surfaces are protected with clean release liners to prevent RNase contamination. HybriSlips remain flat and will not curl, even at high temperatures.
Features and Benefits
- Ready-to-use
- RNase and DNase free
- Hydrophobic
- Disposable
- Will not chip, break or curl
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description L × W × thickness 22 mm × 40 mm × 0.25 mm, pkg of 100 ea | description L × W × thickness 22 mm × 40 mm × 0.25 mm, pkg of 1000 ea | description L × W × thickness 24 mm × 30 mm × 0.25 mm, pkg of 100 ea | description L × W × thickness 22 mm × 22 mm × 0.25 mm, pkg of 1000 ea |
| manufacturer/tradename Grace Bio-Labs 714022 | manufacturer/tradename Grace Bio-Labs 724022 | manufacturer/tradename Grace Bio-Labs 702430 | manufacturer/tradename Grace Bio-Labs 722222 |
| material colorless , polycarbonate | material colorless , polycarbonate | material colorless , polycarbonate | material colorless , polycarbonate |
| sterility non-sterile | sterility non-sterile | sterility - | sterility non-sterile |
| packaging pkg of 100 ea | packaging pkg of 1000 ea | packaging pkg of 100 ea | packaging pkg of 1000 ea |
| L × W × thickness 22 mm × 40 mm × 0.25 mm | L × W × thickness 22 mm × 40 mm × 0.25 mm | L × W × thickness 24 mm × 30 mm × 0.25 mm | L × W × thickness 22 mm × 22 mm × 0.25 mm |
Still not finding the right product?
Explore all of our products under Grace Bio-Labs HybriSlip™ hybridization cover
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?