O této položce
NACRES:
MA.01
UNSPSC Code:
41116155
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimaterial
silicone
cavity size × depth
7-14 mm -4 mm
size
55-76 mm , mold
tip size
-4 mm
color
clear
General description
With cavities for the preparation of microscopy samples. Two variations are available. Maroon molds contain additives for chemical and thermal resistance; they are durable and economical. Clear molds are used where optical clarity is necessary for precise sample alignment; recommended for use with low-temperature UV-cured resins.
Still not finding the right product?
Explore all of our products under Embedding molds
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?