Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
24 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 11 200,00 Kč |
O této položce
11 200,00 Kč
material
polycarbonate bottom (baffled), wide-mouth
sterility
sterile
feature
closure type Vented cap
packaging
pack of 24 ea
manufacturer/tradename
Corning 431682
capacity
5 L
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| feature closure type Vented cap | feature cap, closure type Vented cap | feature cap, closure type Vented cap | feature cap, disposable, Flask style: Erlenmeyer (Fernbach design) |
| material polycarbonate bottom (baffled), wide-mouth | material flat bottom flask, polycarbonate , polypropylene cap, vent cap, wide-mouth , wide-mouth neck | material flat bottom flask, polycarbonate , polypropylene cap, vent cap, wide-mouth , wide-mouth neck | material flat bottom flask, polycarbonate , polypropylene cap (vented, Fernbach design), vent cap, wide-mouth , wide-mouth neck |
| manufacturer/tradename Corning 431682 | manufacturer/tradename Corning 431684 | manufacturer/tradename Corning 431685 | manufacturer/tradename Corning 431252 |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| packaging pack of 24 ea | packaging pack of 1 ea, case of 4 ea | packaging pack of 1 ea, case of 4 ea | packaging pack of 1, case of 4 |
| capacity 5 L | capacity 5 L | capacity 5 L | capacity - |
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?