Přejít k obsahu
Merck

OP64F

Anti-p21WAF1 (58-77) (Ab-1) Mouse mAb (EA10) Fluorescein Conjugate

liquid, clone EA10, Calbiochem®

Synonyma:

Anti-CIP1, Anti-SD11, Anti-p21, Anti-WAF

Přihlásit pro zobrazení organizačních a smluvních cen.

Vybrat velikost


O této položce

UNSPSC Code:
12352203
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli

Název produktu

Anti-p21WAF1 (58-77) (Ab-1) Mouse mAb (EA10) Fluorescein Conjugate, liquid, clone EA10, Calbiochem®

biological source

mouse

antibody product type

primary antibodies

clone

EA10, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

should not react with

mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

Quality Level

Analysis Note

Negative Control
Unstimulated cells, unstimulated skin tissue, or SK-OV-3 cells
Positive Control
Any cell line expressing wild-type p53 (e.g. Hs27 cells or U2OS cells treated with DNA damaging agents) or UV-treated skin or colon tissue

Application

Flow Cytometry (2 µg/ml)

Frozen Sections (5 µg/ml)

Immunoblotting (use Cat. No. OP64)

Immunofluorescence (1-5 µg/ml)

Immunoprecipitation (use Cat. No. OP64)

Paraffin Sections (5 µg/ml, see comments)

Disclaimer

Toxicity: Standard Handling (A)

General description

This product has been discontinued.



Recognizes the ~21 kDa p21WAF1 protein in skin and colon tissue and in cells expressing wild-type p53 (e.g. Hs27 or U205 cells treated with DNA damaging agents).

Purified mouse monoclonal antibody generated by immunizing F1 mice with the specified immunogen and fusing splenocytes with SP2/0 mouse myeloma cells (see application references). Recognizes the ~21 kDa p21WAF1 protein.

Immunogen

Epitope: within amino acids 58-77 of human p21WAF1
Human
full-length, recombinant, human p21WAF1

Other Notes

Agarwal, M.L., et al. 1995. Proc. Natl. Acad. Sci. USA92, 8493.
Chen, Y.Q., et al. 1995. Int. J. Oncology7, 889.
Deng, C., et al. 1995. Cell82, 675.
Waldman, T., et al. 1995. Cancer Res.55, 5187.
Elbendary, A., et al.1994. Cell Growth Diff.5, 1301.
El-Deiry, W.S., et al. 1994 Cancer Res.54, 1169.
Li, R., et al. 1994. Nature371, 534.
Michieli, P., et al. 1994. Cancer Res.54, 3391.
Noda, A., et al.1994. Exp. Cell Res.211, 90.
El-Deiry, W.S., et al.1993. Cell75, 817.
Gu, Y., et al. 1993. Nature366, 707.
Harper, J.W., et al.1993. Cell75, 805.
Xiong, Y., et al.1993. Genes Devel.7, 1572.
Xiong, Y., et al.1993. Nature366, 701.
Xiong, Y., et al.1992. Cell71, 505.
Maximal p21WAF1 expression requires wild type p53 activity. Treatment of U2OS or MCF7 cells with DNA damaging agents (such as doxorubicin at 0.2 µg/ml) induces wild type p53 expression which in turn activates p21WAF1 expression. Serum stimulation of quiescent cells will give low level p21WAF1 expression independent of p53 expression. Untreated cells will express little p21WAF1 and can also be used as a negative control. Cat. No. OP64F was tested in HALT cells induced by incubation at 31°C; FITC-goat anti-mouse IgG (Cat. No. DC13L) was used as a negative control. For staining paraffin sections, no pretreatment is usually needed, but some tissues may stain better after pretreating with heat or pressure cooker. In either paraffin or frozen sections of normal human colon, the non-dividing cells of colonic epithelium will stain positive for p21WAF1 while the proliferating compartment of crypts will not stain. For immunoprecipitation and immunoblotting use Cat. No. OP64. Antibody should be titrated for optimal results in individual systems.

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 50 mM sodium phosphate buffer, 50% glycerol.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Ještě jste nenalezli správný produkt?  

Vyzkoušejte náš produkt Nástroj pro výběr produktů.


Osvědčení o analýze (COA)

Vyhledejte osvědčení Osvědčení o analýze (COA) zadáním čísla šarže/dávky těchto produktů. Čísla šarže a dávky lze nalézt na štítku produktu za slovy „Lot“ nebo „Batch“.

Již tento produkt vlastníte?

Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

Navštívit knihovnu dokumentů

Anca Bene et al.
Biochemical and biophysical research communications, 380(2), 211-217 (2009-01-24)
We have shown previously that in KB-3 (HeLa) cells vinblastine causes downregulation of the CDK inhibitor p21 through a c-Jun regulated pathway. To test the hypothesis that p21 downregulation is necessary to alleviate a protective function, we transfected p21 in
Rajasubramaniam Shanmugam et al.
International journal of cancer, 128(10), 2481-2494 (2010-07-30)
Dimethylaminoparthenolide (DMAPT) is a water soluble parthenolide analog with preclinical activity in hematologic malignancies. Using non-small lung cancer (NSCLC) cell lines (A549 and H522) and an immortalized human bronchial epithelial cell line (BEAS2B) and TCC cell lines (UMUC-3, HT-1197 and
F Noya et al.
Journal of virology, 75(13), 6121-6134 (2001-06-08)
The human papillomavirus (HPV) E7 protein promotes S-phase reentry in a fraction of postmitotic, differentiated keratinocytes. Here we report that these cells contain an inherent mechanism that opposes E7-induced DNA replication. In organotypic raft cultures of primary human keratinocytes, neither

Náš tým vědeckých pracovníků má zkušenosti ve všech oblastech výzkumu, včetně přírodních věd, materiálových věd, chemické syntézy, chromatografie, analytiky a mnoha dalších..

Obraťte se na technický servis