Přejít k obsahu
Merck

NB21

Anti-N-Ethylmaleimide-Sensitive Factor Mouse mAb (NSF-1)

liquid, clone NSF-1, Calbiochem®

Synonyma:

Anti-NSF

Přihlásit pro zobrazení organizačních a smluvních cen.

Vybrat velikost

Změnit zobrazení

O této položce

UNSPSC Code:
12352203
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli


biological source

mouse

Quality Level

antibody product type

primary antibodies

clone

NSF-1, monoclonal

form

liquid

contains

3 mM sodium azide as preservative

species reactivity

human, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

General description

This product has been discontinued.



Recognizes the ~76 kDa NSF-1 protein in human fibroblasts, MDBK, and PC12 cells.

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with mouse myeloma cells. Recognizes the ~76 kDa NSF-1 protein.

Immunogen

Human
recombinant, human N-ethylmaleimide-senstive factor

Application

ELISA (1:2000)

Immunoblotting (1:2000)

Immunoprecipitation (1:500)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 150 mM NaCl, 20 mM sodium phosphate, 50% glycerol, pH 7.5.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Human or rat fibroblasts, MDBK cells, or PC12 cells

Other Notes

Sutton, R.B., et al. 1998. Nature395, 347.
Lemons, P.P., et al. 1997. Blood90, 1490.
Edelmann, L., et al. 1995. EMBO J.14, 224.
Südhof, T.C. 1995. Nature375, 645.
Whiteheart, S.W., et al. 1994. J. Cell Biol.126, 945.
This antibody is suitable for ELISA and immunoprecipitation under both native and denaturing conditions. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Vyzkoušejte náš nástroj Nástroj pro výběr produktů a zúžte své možnosti.


Skladovací třída

10-13 - German Storage Class 10 to 13



Osvědčení o analýze (COA)

Vyhledejte osvědčení Osvědčení o analýze (COA) zadáním čísla šarže/dávky těchto produktů. Čísla šarže a dávky lze nalézt na štítku produktu za slovy „Lot“ nebo „Batch“.

Již tento produkt vlastníte?

Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

Navštívit knihovnu dokumentů



Mark Boltengagen et al.
Nucleic acids research, 44(5), 2145-2159 (2015-11-21)
The incorporation of CENP-A into centromeric chromatin is an essential prerequisite for kinetochore formation. Yet, the molecular mechanisms governing this process are surprisingly divergent in different organisms. While CENP-A loading mechanisms have been studied in some detail in mammals, there
Dakshnamurthy Selvakumar et al.
The Biochemical journal, 474(1), 79-104 (2016-11-09)
Dopamine receptors regulate exocytosis via protein-protein interactions (PPIs) as well as via adenylyl cyclase transduction pathways. Evidence has been obtained for PPIs in inner ear hair cells coupling D1A to soluble N-ethylmaleimide-sensitive factor (NSF) attachment protein receptor (SNARE)-related proteins snapin
Sang H Lee et al.
Nature communications, 9(1), 3434-3434 (2018-08-26)
Secreted Wnts play crucial roles in synaptogenesis and synapse maintenance, but endogenous factors promoting synapse elimination in central neurons remain unknown. Here we show that proline-rich 7 (PRR7) induces specific removal of excitatory synapses and acts as a Wnt inhibitor.