Přejít k obsahu
Merck

MABN279

Anti-Beta-mannosidase Antibody, clone 1B3.1

clone 1B3.1, from mouse

Synonyma:

Beta-mannosidase, Lysosomal beta A mannosidase, Mannanase, Mannase

Přihlásit pro zobrazení organizačních a smluvních cen.

Vybrat velikost

Změnit zobrazení

O této položce

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
1B3.1, monoclonal
Application:
IHC, WB
Citations:
-
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1B3.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MANBA(4126)

General description

Beta-mannosidase, also known as Lysosomal beta A mannosidase or Mannanase, or Mannase, and encoded by the gene MANBA/MANB1, is an exoglycosidase that cleaves the single beta-linked mannose residue from the non-reducing end of all N-linked glycoprotein oligosaccharides. Beta-mannosidase is localized to the lysosome. Beta-mannosidase is expressed in many cell types including fibroblasts, neutrophils and even neurons and glia. Defects in Beta-mannosidase may be associated with a mild disorder that affects peripheral and central nervous system myelin health and is called Mannosidoisis beta A. It is a lysosomal storage disorder that results in a mixture of clinical features from mental retardation, hearing loss and speech activities.
~80 kDa observed. The calculated molecular weight of this protein is 100 kDa However, this protein undergoes post-translational modification, and may be observed at ~110 kDa, ~100 kDa, ~91 kDa, ~86 kDa, ~84 kDa, and ~75 kDa (Sopher, B. L., et al. (1993). Biochem J. 289(Pt 2):343-347.).

Immunogen

KLH-conjugated linear peptide corresponding to human Beta-mannosidase.

Application

Anti-Beta-mannosidase Antibody, clone 1B3.1 is a highly specific mouse monoclonal antibody, that targets Mannosidase & has been tested in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected Beta-mannosidase in human cerebral cortex tissue.
Research Category
Neuroscience
Research Sub Category
Signaling Neuroscience

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected Beta-mannosidase in 10 µg of HepG2 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Vyzkoušejte náš nástroj Nástroj pro výběr produktů a zúžte své možnosti.


Skladovací třída

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Osvědčení o analýze (COA)

Vyhledejte osvědčení Osvědčení o analýze (COA) zadáním čísla šarže/dávky těchto produktů. Čísla šarže a dávky lze nalézt na štítku produktu za slovy „Lot“ nebo „Batch“.

Již tento produkt vlastníte?

Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

Navštívit knihovnu dokumentů



Globální číslo obchodní položky

Skladová položkaGTIN
MABN27904053252933998