Přejít k obsahu
Merck

HTS073M

ChemiSCREEN Recombinant Human β2 Adrenoceptor

Přihlásit pro zobrazení organizačních a smluvních cen.

Vybrat velikost

Změnit zobrazení

O této položce

eCl@ss:
32161000
UNSPSC Code:
41123201
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli


Quality Level

description

GPCR Class A, protein target beta 2

technique(s)

radioligand binding assay (RLBA): suitable

UniProt accession no.

Gene Information

human ... ADRBK2(157)

General description

FUNCTION: SwissProt: P35626 # Specifically phosphorylates the agonist-occupied form of the beta-adrenergic and closely related receptors.
SIZE: 688 amino acids; 79710 Da
SIMILARITY: SwissProt: P35626 ## Belongs to the protein kinase superfamily. AGC Ser/Thr protein kinase family. GPRK subfamily. & Contains 1 AGC-kinase C-terminal domain. & Contains 1 PH domain. & Contains 1 protein kinase domain. & Contains 1 RGS domain.

Application

Radioligand binding assay, and GTPγS binding.

Physical form

One package contains enough membranes for at least 200 assays (units), where an unit is the amount of membrane that will yield greater than 40-fold signal:background with 125I-labeled (-)Iodocyanopindolol at 0.5 nM. Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives. Packaging method: Membranes protein were adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80oC.

Preparation Note

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Analysis Note

Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 50 mM Hepes, pH 7.4, 5 mM MgCl2, 1 mM CaCl2, filtered and stored at 4°C
Radioligand: [125I]-(-) Iodocyanopindolol (Perkin Elmer # NEX189)
Wash Buffer: 50 mM Hepes, pH 7.4, 500 mM NaCl, filtered and stored at 4°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Skladovací třída

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Osvědčení o analýze (COA)

Vyhledejte osvědčení Osvědčení o analýze (COA) zadáním čísla šarže/dávky těchto produktů. Čísla šarže a dávky lze nalézt na štítku produktu za slovy „Lot“ nebo „Batch“.

Již tento produkt vlastníte?

Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

Navštívit knihovnu dokumentů