Přejít k obsahu
Merck

650212

ProteoExtract® All-in-One Trypsin Digestion Kit

Přihlásit pro zobrazení organizačních a smluvních cen.

Vybrat velikost


O této položce

UNSPSC Code:
41106500
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli

usage

sufficient for 100 digest(s)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

An optimized kit for the digestion of proteins by trypsin either in-gel, in-solution or in-suspension. Digested samples are suitable for analysis on mass spectroscopy, HPLC, or electrophoresis.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Sequencing Grade Trypsin, Extraction Buffers, Solvents, Digest Buffer, Reducing Agent, Blocking Agent, Wash Buffer, and a user protocol.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PROTEOEXTRACT is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Osvědčení o analýze (COA)

Vyhledejte osvědčení Osvědčení o analýze (COA) zadáním čísla šarže/dávky těchto produktů. Čísla šarže a dávky lze nalézt na štítku produktu za slovy „Lot“ nebo „Batch“.

Již tento produkt vlastníte?

Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

Navštívit knihovnu dokumentů

Hardeep K Gumber et al.
Journal of cell science, 132(3) (2019-01-20)
The linker of nucleoskeleton to cytoskeleton (LINC) complex is an essential multi-protein structure spanning the nuclear envelope. It connects the cytoplasm to the nucleoplasm, functions to maintain nuclear shape and architecture and regulates chromosome dynamics during cell division. Knowledge of
Masataka Kawai et al.
Biophysical journal, 112(8), 1726-1736 (2017-04-27)
The cardiac troponin C (TnC)-A8V mutation is associated with hypertrophic and restrictive cardiomyopathy (HCM and RCM) in human and mice. The residue affected lies in the N-helix, a region known to affect Ca2+-binding affinity to the N-terminal domain. Here we
Maicon Landim-Vieira et al.
eLife, 11 (2022-05-04)
Phosphorylation and acetylation of sarcomeric proteins are important for fine-tuning myocardial contractility. Here, we used bottom-up proteomics and label-free quantification to identify novel post-translational modifications (PTMs) on β-myosin heavy chain (β-MHC) in normal and failing human heart tissues. We report
Su Su et al.
Cell reports, 25(11), 3215-3228 (2018-12-13)
Molecular- and cellular-based therapies have the potential to reduce obesity-associated disease. In response to cold, beige adipocytes form in subcutaneous white adipose tissue and convert energy stored in metabolic substrates to heat, making them an attractive therapeutic target. We developed a
Meenakshi Agarwal et al.
Cells, 7(12) (2018-12-14)
Two Burkholderia spp. (strains SRS-25 and SRS-46) were isolated from high concentrations of uranium (U) from the U.S. Department of Energy (DOE)-managed Savannah River Site (SRS). SRS contains soil gradients that remain co-contaminated by heavy metals from previous nuclear weapons

Náš tým vědeckých pracovníků má zkušenosti ve všech oblastech výzkumu, včetně přírodních věd, materiálových věd, chemické syntézy, chromatografie, analytiky a mnoha dalších..

Obraťte se na technický servis