Přejít k obsahu
Merck

616377

Toxin B, Clostridium difficile

A high molecular weight glucosyltransferase that inhibits small GTPases, such as Rho, Rac, and Cdc42, by glucosylation of a threonine residue.

Synonyma:

Cytotoxin

Přihlásit pro zobrazení organizačních a smluvních cen.

Vybrat velikost


O této položce

UNSPSC Code:
12352200
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli

Název produktu

Toxin B, Clostridium difficile, A high molecular weight glucosyltransferase that inhibits small GTPases, such as Rho, Rac, and Cdc42, by glucosylation of a threonine residue.

form

lyophilized

manufacturer/tradename

Calbiochem®

availability

not available in, Canada

storage condition

do not freeze

solubility

sterile distilled water: soluble

storage temp.

2-8°C

Quality Level

Analysis Note

major band at >200 kDa

Biochem/physiol Actions

Cytotoxic as determined in a cell rounding assay using human foreskin cells.
Primary Target
Small GTPases, such as Rho, Rac, and Cdc42
Product does not compete with ATP.

Disclaimer

Toxicity: Irritant (B)

General description

A high molecular weight glucosyltransferase that inhibits small GTPases, such as Rho, Rac, and Cdc42, by glucosylation of a threonine residue.
A high molecular weight glucosyltransferase that inhibits small GTPases, such as Rho, Rac, and Cdc42, by glucosylation of a threonine residue. This results in the shutdown of signal transduction cascades, leading to depolymerization of the cytoskeleton, gene transcription of a number of stress-activated protein kinases (SAPKs), and a decrease in the synthesis of phosphatidylinositiol 4,5-bisphosphate. May also block serotonin release, and stimulate the production of phospholipases C and D. Due to its ability to affect depolymerization of the cytoskeleton, toxin B has been shown to inhibit biphasic muscle contraction. Unlike Exoenzyme C3 (Cat. No. 341208), this toxin does not require assistance to permeabilize cells.

Other Notes

Mackay, D.J. and Hall, A. 1998. J. Biol. Chem. 273, 20685.
Prepens, U., et al. 1996. J. Biol. Chem. 271, 7324.
Schmidt, M., et al. 1996. Eur. J. Biochem. 240, 707.
Wilkins, T.D. and Lyerly, D.M. 1996. Trends Microbiol. 4, 49.

Physical form

Lyophilized from 50 mM Tris, 50 mM NaCl, 0.1% trehalose, pH 7.5.

Preparation Note

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 2 weeks at 4°C. Toxin B activity will decrease after 2 weeks at 4°C following reconstitution.
Please see vial label for lot-specific reconstitution volume.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal

Skladovací třída

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3


Osvědčení o analýze (COA)

Vyhledejte osvědčení Osvědčení o analýze (COA) zadáním čísla šarže/dávky těchto produktů. Čísla šarže a dávky lze nalézt na štítku produktu za slovy „Lot“ nebo „Batch“.

Již tento produkt vlastníte?

Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

Navštívit knihovnu dokumentů

Náš tým vědeckých pracovníků má zkušenosti ve všech oblastech výzkumu, včetně přírodních věd, materiálových věd, chemické syntézy, chromatografie, analytiky a mnoha dalších..

Obraťte se na technický servis