Přejít k obsahu
Merck

407611

Interleukin-1α, Human, Recombinant, E. coli

Přihlásit pro zobrazení organizačních a smluvních cen.

Vybrat velikost


O této položce

NACRES:
NA.77
UNSPSC Code:
12352202
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli

assay

≥97% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1.0 EU/μg Endotoxin (EU/μg IL-1α)

shipped in

ambient

storage temp.

−70°C

Quality Level

Biochem/physiol Actions

ED₅₀ = 3-7 pg/ml as measured in a cell proliferation assay.

Disclaimer

Toxicity: Standard Handling (A)

General description

Recombinant, human interleukin-1α expressed in E. coli. A 159 amino acid polypeptide derived from mature IL-1α. Synthesized in vivo as a 271 amino acid residue (31 kDa) precursor that is subsequently cleaved into the mature polypeptide. Shows ~25% amino acid sequence identity with IL-1β, a distinct gene product sharing the same cell surface receptors as IL-1α. Produced by activated macrophages and many other cell types, the IL-1 proteins mediate a variety of effects on the differentiation and function of many cell types involved in inflammatory and immune responses. Potent stimulator of bone resorption and stimulator of nuclear phospholipase C. Induces nitric oxide synthase. Effects include stimulation of T-cell, B-cell, neutrophil, and fibroblast proliferation; induction of prostaglandin, leukotriene, and acute-phase hepatic protein synthesis; induction of proteinases in synovial fibroblasts and articular chondrocytes. IL-1 is also known to induce fever, slow-wave sleep, and bone resorption. 1 µg is equivalent to 1.6 x 105 WHO units.

Other Notes

Tsan, M.F., and White, J.E. 1994. Am. J. Physiol. 266, L316.
Kilbourn, R.G., et al. 1992. J. Natl. Cancer Inst. 84, 1008.
Dinarello, C.A., et al. 1991. Blood77, 1627.
Casagli, M.C., et al. 1989. Biochem. Biophys. Res. Commun.162, 357.
Symons, J.A., et al. 1987. In Lymphokines and Interferons, A Practical Approach (Clemens, M.J., et al. Eds.) Oxford IRL Press, p. 272.
March, C.J., et al. 1985. Nature315, 641.
Kampschmidt, R.F. 1984. J. Leuk. Biol.36, 341.

Physical form

Lyophilized from sterile-filtered 20 mM Tris-HCl, 50 µg BSA/µg IL-1α, pH 8.3.

Preparation Note

Following reconstitution, aliquot and freeze (-70°C) for long term storage or refrigerate (4°C) for short term storage. Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -70°C.
Reconstitute with sterile PBS containing ≥0.1% HSA or BSA.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Skladovací třída

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Osvědčení o analýze (COA)

Vyhledejte osvědčení Osvědčení o analýze (COA) zadáním čísla šarže/dávky těchto produktů. Čísla šarže a dávky lze nalézt na štítku produktu za slovy „Lot“ nebo „Batch“.

Již tento produkt vlastníte?

Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

Navštívit knihovnu dokumentů

M C Casagli et al.
Biochemical and biophysical research communications, 162(1), 357-363 (1989-07-14)
Human recombinant interleukin 1 beta produced in Escherichia coli and in Saccharomyces cerevisiae was purified to homogeneity by a combination of ion exchange, gel filtration and hydroxylapatite column chromatography. The two proteins, both expressed in the mature form, differ in
C A Dinarello
Blood, 77(8), 1627-1652 (1991-04-15)
The polypeptide cytokine interleukin-1 (IL-1) affects nearly every tissue and organ system. IL-1 is the prototype of the pro-inflammatory cytokines in that it induces the expression of a variety of genes and the synthesis of several proteins that, in turn
The numerous postulated biological manifestations of interleukin-1.
R F Kampschmidt
Journal of leukocyte biology, 36(3), 341-355 (1984-09-01)
R G Kilbourn et al.
Journal of the National Cancer Institute, 84(13), 1008-1016 (1992-07-01)
Interleukin-1-alpha (IL-1) is a cytokine with potentially therapeutic immunoproliferative and tumoricidal activities. Preliminary clinical studies suggest that use of IL-1 may be restricted by dose-limiting hypotension. The purpose of this study was to investigate the role of nitric oxide (NO.)
Interleukin-1alpha induced protection against pulmonary oxygen toxicity.
M F Tsan et al.
The American journal of physiology, 266(3 Pt 1), L316-L317 (1994-03-01)

Náš tým vědeckých pracovníků má zkušenosti ve všech oblastech výzkumu, včetně přírodních věd, materiálových věd, chemické syntézy, chromatografie, analytiky a mnoha dalších..

Obraťte se na technický servis