Přejít k obsahu
Merck
40-095
BioTek® 405 LS Magnetic/Vacuum Filtration 96- well Washer
Synonyma:
Vacuum filtration system
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
O této položce
eCl@ss:
42029053
UNSPSC Code:
41116010
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
product line
Milliplex®
technique(s)
multiplexing: suitable
General description
The BioTek® Magnetic/Vacuum filtration 96 well washer is based on the industry standard in microplate washing with superior performance and reliability. This fifth generation product offers processing of 96- well microplates. The BioTek® Magnetic/Vacuum filtration 96 well washer provides excellent performance for ELISA and cell-based assays. Programming the BioTek® Magnetic/Vacuum filtration 96 well washer is easy through its simple interface. For those who prefer the advantages of running instruments via computer control or require 21 CFR Part 11 compliance, the BioTek® Magnetic/Vacuum filtration 96 well washer is controlled with BioTek®′s Liquid Handling Control Software.
For washing microsphere based assays, the BioTek® Magnetic/Vacuum filtration 96 well washer is equipped with accessory modules for both biomagnetic separation and vacuum filtration. This allows for full plate washing of magnetic and polystyrene beads used in a growing number of multiplex assays and bead-based ELISAs, such as those developed on the Luminex® xMAP® technology platform. Developed with leaders in genotyping, gene expression and protein assays, the BioTek® Magnetic/Vacuum filtration 96 well washer biomagnetic separation incorporates high-energy neodymium iron boron magnets for rapid separation of micrometer and nanometer beads with superior retention. The BioTek® Magnetic/Vacuum filtration 96 well washer vacuum filtration feature automates the wash steps of filter bottom plates with five different vacuum levels for optimal performance of filter pore sizes ranging from 0.45 µm to 1.2 µm. The BioTek® Magnetic/Vacuum filtration 96 well washer is Luminex® xMAP® approved. The BioTek® Magnetic/Vacuum filtration 96 well washer is an ideal choice with unique features to lower background absorbance values and increase the signal-to-noise ratios.
Application
The BioTek® Magnetic/Vacuum filtration 96 well washer provides excellent performance for ELISA & cell-based assays.
For use with MILLIPLEX® MAP assays, see product protocol for specific use instructions
Other Notes
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial
Legal Information
BioTek is a registered trademark of BioTek/Fischer
Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp
Osvědčení o analýze (COA)
Vyhledejte osvědčení Osvědčení o analýze (COA) zadáním čísla šarže/dávky těchto produktů. Čísla šarže a dávky lze nalézt na štítku produktu za slovy „Lot“ nebo „Batch“.
Již tento produkt vlastníte?
Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.
Navštívit knihovnu dokumentů
Náš tým vědeckých pracovníků má zkušenosti ve všech oblastech výzkumu, včetně přírodních věd, materiálových věd, chemické syntézy, chromatografie, analytiky a mnoha dalších..
Obraťte se na technický servis