Přejít k obsahu
Merck

345818

RapidStep ECL Reagent

This RapidStep ECL Reagent is validated for use in Immunoblotting for the detection of RapidStep ECL Reagent.

Přihlásit pro zobrazení organizačních a smluvních cen.

Vybrat velikost

Změnit zobrazení

O této položce

UNSPSC Code:
12352200
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli


form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

shipped in

wet ice

storage temp.

2-8°C

General description

A single component reagent for chemiluminescent detection of immobilized proteins, labeled with horseradish peroxidase, by immunoblotting. Provides a bright, stable signal with low signal:background. Does not require mixing of substrate and enhancer solutions. Volume is sufficient for covering up to 2000 cm2 of membrane.
A single component reagent for chemiluminescent detection of immobilized proteins, labeled with horseradish peroxidase, by immunoblotting. Volume supplied is sufficient for covering up to 2000 cm2 of membrane.
RapidStep ECL is a 1 STEP chemiluminescent substrate reagent for detection of horseradish peroxidase (HRP) on immunoblots. It is a convenient, cost-effective, and ready-to-use reagent that provides a superior, longer-lasting signal than traditional ECL reagents.

RapidStep contains luminol and enhancer combined in one solution and is ready-to-use without mixing. A spray bottle is used to apply the solution directly onto the membrane.
This RapidStep ECL Reagent is validated for use in Immunoblotting for the detection of RapidStep ECL Reagent.

Application

Immunoblotting

Other Notes

To achieve an optimal signal:noise ratio, it is necessary to optimize the concentration of primary and secondary antibodies.

Protocol for Use

1. Remove the membrane containing immobilized proteins from the last wash and place on a flat surface, protein side facing up. Do not allow the membrane to dry

2. Spray RapidStep ECL Reagent on the surface of the membrane to sufficiently cover the entire membrane. Gently shake the container to ensure the membrane is evenly covered. Generally 2 sprays are sufficient to cover a 5 cm x 5 cm area.

3. Incubate the membrane for 1 min at room temperature without agitation. Proteins with low specificity or low reactivity with the primary antibody may require longer incubation times.

4. Drain the excess RapidStep ECL Reagent by holding the membrane vertically and touch the edge of the membrane against absorbent paper. Failure to drain sufficiently may lead to high background.

5. Wrap the membrane in plastic wrap and gently smooth out air pockets.

6. Place the membrane in a film cassette, protein side facing up.

7. Place X-ray film on top of the membrane, close the cassette and expose for an appropriate period of time. Optimal exposure times should be determined for each experiment.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Skladovací třída

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Osvědčení o analýze (COA)

Vyhledejte osvědčení Osvědčení o analýze (COA) zadáním čísla šarže/dávky těchto produktů. Čísla šarže a dávky lze nalézt na štítku produktu za slovy „Lot“ nebo „Batch“.

Již tento produkt vlastníte?

Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

Navštívit knihovnu dokumentů



Yuan-Ya Jing et al.
Cell death and differentiation, 27(5), 1660-1676 (2019-11-07)
Cancer cells reprogram their energy metabolic system from the mitochondrial oxidative phosphorylation (OXPHOS) pathway to a glucose-dependent aerobic glycolysis pathway. This metabolic reprogramming phenomenon is known as the Warburg effect, a significant hallmark of cancer. However, the detailed mechanisms underlying
Xinyu Zhuang et al.
Experimental and therapeutic medicine, 14(1), 515-524 (2017-07-05)
The mitochondria are highly dynamic organelles, carefully maintaining network homeostasis by regulating mitochondrial fusion and fission. Mitochondrial dynamics are involved in the regulation of a variety of pathophysiological processes, including cell proliferation. Oxidative stress serves an important role in the
Kui Ge et al.
Frontiers in plant science, 8, 1150-1150 (2017-07-18)
Abscisic acid (ABA) transporters are essential for the transport of ABA from its sites of synthesis to its multiple sites of action within plants and are key players in plant stress responses. Despite their importance, there is limited information on