Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohliQuality Level
form
lyophilized
specific activity
≥200 collagenase units/mg dry wt
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, desiccated
solubility
aqueous buffer: soluble
foreign activity
Non-specific proteolytic ≥50 fluorescence units/mg
shipped in
ambient
storage temp.
2-8°C
General description
Crude collagenase preparation that contains higher clostripain and tryptic activity than Type I. Useful in tissue dissociation studies in which a combination of collagenase and high proteolytic activity is required.
Crude collagenase solid prepared to contain higher clostripain and tryptic activity than Type I. Useful in tissue dissociation studies in which a combination of collagenase and high proteolytic activity is required. Has an optimal pH of 7.0-8.0.
Preparation Note
Unstable in solution; reconstitute just prior to use.
Other Notes
Keller, S., and Mandl, I. 1963. Arch. Biochem. Biophys.101, 81.
One unit is defined as the amount of enzyme that will release 1.0 nmol Leu equivalent from collagen per min at 37°C, pH 7.2.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Harmful (C)
Skladovací třída
10-13 - German Storage Class 10 to 13
Osvědčení o analýze (COA)
Vyhledejte osvědčení Osvědčení o analýze (COA) zadáním čísla šarže/dávky těchto produktů. Čísla šarže a dávky lze nalézt na štítku produktu za slovy „Lot“ nebo „Batch“.
Již tento produkt vlastníte?
Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.
Huai-Chun Yang et al.
World journal of stem cells, 12(2), 152-167 (2020-03-19)
Human-derived mesenchymal stromal cells have been shown to improve cognitive function following experimental stroke. The activity of exosomes has been verified to be comparable to the therapeutic effects of mesenchymal stromal cells. However, the effects of exosomes derived from human
Seungil Kim et al.
SLAS discovery : advancing life sciences R & D, 25(7), 744-754 (2020-05-01)
3D cell culture models have been developed to better mimic the physiological environments that exist in human diseases. As such, these models are advantageous over traditional 2D cultures for screening drug compounds. However, the practicalities of transitioning from 2D to