Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 2 900,00 Kč |
O této položce
2 900,00 Kč
material
borosilicate glass flask, pear-shaped , single-neck
joint
joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F909100
capacity
100 mL
General description
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description capacity 100 mL, joint: ST/NS 19/22 | description capacity 100 mL, joint: ST/NS 19/22 | description capacity 100 mL, joint: ST/NS 19/22 | description capacity 100 mL, joint: ST/NS 14/20 |
| manufacturer/tradename Synthware F909100 | manufacturer/tradename Synthware F909100G | manufacturer/tradename Synthware F909100H | manufacturer/tradename Synthware F901100H |
| capacity 100 mL | capacity 100 mL | capacity 100 mL | capacity 100 mL |
| material borosilicate glass flask, single-neck , pear-shaped | material single-neck , pear-shaped , borosilicate glass flask | material borosilicate glass flask, pear-shaped , single-neck | material borosilicate glass flask, single-neck , pear-shaped |
| joint joint: ST/NS19/22 | joint joint: ST/NS19/22 | joint joint: ST/NS19/22 | joint joint: ST/NS14/20 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ pear-shaped Schlenk flask with stopcock
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?