Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 1 500,00 Kč |
O této položce
1 500,00 Kč
material
borosilicate glass flask, round bottom , single-neck
joint
joint: ST/NS35/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F527000
capacity
1,000 mL , 1000 mL , 1000 mL
General description
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description 1000 mL, joint: ST/NS 35/20 | description 1000 mL, joint: ST/NS 24/40 | description 1000 mL, joint: ST/NS 29/42 | description 100 mL, joint: ST/NS 24/40 |
| manufacturer/tradename Synthware F527000 | manufacturer/tradename Synthware F304000 | manufacturer/tradename Synthware F30291L | manufacturer/tradename Synthware F304100 |
| capacity 1000 mL , 1,000 mL , 1000 mL | capacity 1,000 mL , 1000 mL , 1000 mL | capacity 1000 mL , 1000 mL, 1,000 mL | capacity 100 mL, 100 mL |
| material borosilicate glass flask, single-neck , round bottom | material round bottom , single-neck , borosilicate glass flask | material borosilicate glass flask, single-neck , round bottom | material borosilicate glass flask, single-neck , round bottom |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| joint joint: ST/NS35/20 | joint joint: ST/NS24/40 | joint joint: ST/NS29/42 | joint joint: ST/NS24/40 |
Still not finding the right product?
Explore all of our products under Synthware™ single-neck round-bottom flask
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?