Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 3 140,00 Kč |
O této položce
3 140,00 Kč
material
borosilicate glass flask, round bottom , three-neck (vertical side necks)
joint
center joint: ST/NS24/40, side joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F444250
capacity
250 mL, 250 mL
General description
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description 250 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40 | description 250 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20 | description 250 mL, center joint: ST/NS 24/40, side joint: ST/NS 19/22 | description 2000 mL, center joint: ST/NS 29/42, side joint: ST/NS 29/42 |
| capacity 250 mL , 250 mL | capacity 250 mL , 250 mL | capacity 250 mL , 250 mL | capacity 2000 mL, 2000 mL , 2,000 mL |
| manufacturer/tradename Synthware F444250 | manufacturer/tradename Synthware F441250 | manufacturer/tradename Synthware F442250 | manufacturer/tradename Synthware F44292L |
| joint center joint: ST/NS24/40, side joint: ST/NS24/40 | joint center joint: ST/NS14/20, side joint: ST/NS14/20 | joint center joint: ST/NS24/40, side joint: ST/NS19/22 | joint center joint: ST/NS29/42, side joint: ST/NS29/42 |
| material three-neck (vertical side necks), round bottom , borosilicate glass flask | material borosilicate glass flask, three-neck (vertical side necks), round bottom | material three-neck (vertical side necks), borosilicate glass flask, round bottom | material round bottom , borosilicate glass flask, three-neck (vertical side necks) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ three neck round bottom flask with vertical side necks
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?