Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 2 310,00 Kč |
O této položce
2 310,00 Kč
material
borosilicate glass flask, round bottom , three-neck (angled side necks)
joint
center joint: ST/NS24/40, side joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F434150
capacity
50 mL, 50 mL
General description
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description 50 mL, center joint: ST/NS 24/40, side joint: ST/NS 14/20 | description 50 mL, center joint: ST/NS 19/22, side joint: ST/NS 14/20 | description 50 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40 | description 50 mL, center joint: ST/NS 24/40, side joint: ST/NS 19/22 |
| manufacturer/tradename Synthware F434150 | manufacturer/tradename Synthware F439150 | manufacturer/tradename Synthware F434450 | manufacturer/tradename Synthware F434950 |
| capacity 50 mL , 50 mL | capacity 50 mL , 50 mL | capacity 50 mL , 50 mL | capacity 50 mL , 50 mL |
| material borosilicate glass flask, three-neck (angled side necks), round bottom | material borosilicate glass flask, round bottom , three-neck (angled side necks) | material borosilicate glass flask, round bottom , three-neck (angled side necks) | material borosilicate glass flask, round bottom , three-neck (angled side necks) |
| joint side joint: ST/NS14/20, center joint: ST/NS24/40 | joint center joint: ST/NS19/22, side joint: ST/NS14/20 | joint center joint: ST/NS24/40, side joint: ST/NS24/40 | joint center joint: ST/NS24/40, side joint: ST/NS19/22 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ three neck round bottom flask with angled side necks
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?