Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 1 680,00 Kč |
O této položce
1 680,00 Kč
material
borosilicate glass flask, round bottom , two-neck (angled side neck)
joint
center joint: ST/NS24/40, side joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F414450
capacity
50 mL, 50 mL
General description
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description 50 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40 | description 50 mL, center joint: ST/NS 24/40, side joint: ST/NS 14/20 | description 50 mL, center joint: ST/NS 19/22, side joint: ST/NS 19/22 | description 50 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20 |
| capacity 50 mL , 50 mL | capacity 50 mL, 50 mL | capacity 50 mL, 50 mL | capacity 50 mL, 50 mL |
| manufacturer/tradename Synthware F414450 | manufacturer/tradename Synthware F414150 | manufacturer/tradename Synthware F419950 | manufacturer/tradename Synthware F411150 |
| material borosilicate glass flask, two-neck (angled side neck), round bottom | material borosilicate glass flask, round bottom , two-neck (angled side neck) | material round bottom , borosilicate glass flask, two-neck (angled side neck) | material borosilicate glass flask, two-neck (angled side neck), round bottom |
| joint center joint: ST/NS24/40, side joint: ST/NS24/40 | joint center joint: ST/NS24/40, side joint: ST/NS14/20 | joint center joint: ST/NS19/22, side joint: ST/NS19/22 | joint center joint: ST/NS14/20, side joint: ST/NS14/20 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ two neck round bottom flask with angled side neck
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?