Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 3 310,00 Kč |
O této položce
3 310,00 Kč
material
borosilicate glass flask, round bottom , single-neck
joint
joint: ST/NS29/42
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F30292L
capacity
2,000 mL , 2000 mL , 2000 mL
General description
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description 2000 mL, joint: ST/NS 29/42 | description 2000 mL, joint: ST/NS 24/40 | description 2000 mL, joint: ST/NS 45/50 | description 200 mL, joint: ST/NS 24/40 |
| manufacturer/tradename Synthware F30292L | manufacturer/tradename Synthware F30242L | manufacturer/tradename Synthware F30452L | manufacturer/tradename Synthware F304200 |
| capacity 2,000 mL , 2000 mL , 2000 mL | capacity 2,000 mL , 2000 mL, 2000 mL | capacity 2000 mL , 2,000 mL , 2000 mL | capacity 200 mL , 200 mL |
| material round bottom , single-neck , borosilicate glass flask | material borosilicate glass flask, round bottom , single-neck | material single-neck , round bottom , borosilicate glass flask | material round bottom , single-neck , borosilicate glass flask |
| joint joint: ST/NS29/42 | joint joint: ST/NS24/40 | joint joint: ST/NS45/50 | joint joint: ST/NS24/40 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ single-neck round-bottom flask
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?