Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 638,00 Kč |
O této položce
638,00 Kč
material
borosilicate glass flask, round bottom , single-neck
joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F301100
capacity
100 mL, 100 mL
General description
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| capacity 100 mL , 100 mL | capacity 100 mL , 100 mL | capacity 100 mL , 100 mL | capacity 100 mL , 100 mL |
| manufacturer/tradename Synthware F301100 | manufacturer/tradename Synthware F308100 | manufacturer/tradename Synthware F309100 | manufacturer/tradename Synthware F304100 |
| joint joint: ST/NS14/20 | joint joint: ST/NS29/42 | joint joint: ST/NS19/22 | joint joint: ST/NS24/40 |
| material borosilicate glass flask, single-neck , round bottom | material borosilicate glass flask, round bottom , single-neck | material borosilicate glass flask, round bottom , single-neck | material borosilicate glass flask, round bottom , single-neck |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ single-neck round-bottom flask
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?