O této položce
NACRES:
NB.03
UNSPSC Code:
42142600
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimaterial
Gastight syringe
feature
autoclavable, model 1701 RN, no needle
manufacturer/tradename
Hamilton
parameter
10-115 °C temp. range (50-239 °F), 1000 psig pressure
I.D. × O.D.
0.460 mm × 6.60 mm
volume
10 μL
General description
The needles on Thin Layer Chromatography (TLC) syringes are coated from the point for 3/4” with a PTFE-coating. The coating reduces the surface tension between the needle and the liquid making it ideal for reproducible sample spotting. The TLC syringe is available as a cemented needle (SNTLC) or as a replacement needle to be installed into a standard removable needle (RN) syringe. To find the appropriate no needle syringe select general syringes and filter by termination and gauge. Choose the “RN” and “No Needle Included” options to see all syringes that can receive TLC needles. To find the appropriate TLC needle select the TLC option under “Needles” in the main product menu.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Still not finding the right product?
Explore all of our products under Hamilton® TLC syringe
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?