Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 5 330,00 Kč |
O této položce
5 330,00 Kč
material
Gastight syringe
feature
autoclavable: no, model 1005 TLL-SAL, no needle
manufacturer/tradename
Hamilton
parameter
20-50 °C temp. range (68-122 °F), 200 psig pressure
volume
5 mL
General description
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description model, 1005 TLL-SAL, no needle, volume 5 mL | description model, 1725 TLL-SAL, no needle, volume 250 μL | description model, 1750 TLL-SAL, no needle, volume 500 μL | description for CTC/LEAP PAL LC Autosampler, model, 1750 FN CTC, fixed needle, volume × L 500 μL × 51 mm, needle gauge 22 |
| manufacturer/tradename Hamilton | manufacturer/tradename Hamilton | manufacturer/tradename Hamilton | manufacturer/tradename Hamilton |
| feature autoclavable: no, model 1005 TLL-SAL, no needle | feature model 1725 TLL-SAL, no needle, autoclavable: no | feature autoclavable: no, model 1750 TLL-SAL, no needle | feature autoclavable: no, model 1750 FN CTC, fixed needle |
| volume 5 mL | volume 250 μL | volume 500 μL | volume - |
| material Gastight syringe | material Gastight syringe | material Gastight syringe | material Gastight syringe |
| parameter 200 psig pressure | parameter 500 psig pressure, 20-50 °C temp. range (68-122 °F) | parameter 700 psig pressure, 20-50 °C temp. range (68-122 °F) | parameter 20-50 °C temp. range (68-122 °F), 700 psig pressure |
Still not finding the right product?
Explore all of our products under Hamilton® SaltLine reagent syringe
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?