Změnit zobrazení
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
10 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 3 340,00 Kč |
O této položce
NACRES:
NB.43
UNSPSC Code:
41121803
Material:
borosilicate glass 3.3
Feature:
with graduation
3 340,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimaterial
borosilicate glass 3.3
feature
with graduation
manufacturer/tradename
BRAND 91241
volume
400 mL
General description
The BRAND® Glass Beaker with Spout is expertly crafted from Boro 3.3 glass, making it a reliable choice for laboratory applications. Designed in accordance with DIN EN ISO 3819 standards, this low-form beaker features clear graduation markings for precise measurement of liquids. The integrated spout allows for easy pouring, minimizing spills and enhancing accuracy during liquid transfer.
Other Notes
Browse all glass beakers
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
1 of 1
Tato položka | |||
|---|---|---|---|
| material borosilicate glass 3.3 | material borosilicate glass 3.3 | material borosilicate glass 3.3 | material borosilicate glass 3.3 |
| feature with graduation | feature with graduation | feature with graduation | feature with graduation |
| manufacturer/tradename BRAND 91241 | manufacturer/tradename BRAND 91253 | manufacturer/tradename BRAND 91248 | manufacturer/tradename BRAND 91236 |
| volume 400 mL | volume 800 mL | volume 600 mL | volume 250 mL |
Still not finding the right product?
Explore all of our products under BRAND® glass beaker with spout, low form
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?