Změnit zobrazení
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 709,00 Kč |
O této položce
NACRES:
NB.01
UNSPSC Code:
41121800
Material:
FEP
709,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimaterial
FEP
packaging
pack of 1 ea
manufacturer/tradename
BRAND 704209
General description
Dispense accurate volumes directly from drums and bulk refills.The Dispensette® can be mounted on a wall, a ring stand or on lab furniture.A filter in the drum adapter minimizes risk of contaminating highpurity reagents when air is drawn in for pressure compensation after dispensing.A quick-release connector with integrated valves allows quick changing of the bulk container.The remote dispensing system allows storage of the drum up to 10 meters (30 feet) away from the Dispensette®.The max. delivery height is approximately 1.2 m.Operating Exclusions: Never use the remote dispensing system: 1. with recirculation valve. It has to be removed before use! 2. for pressurized vessels 3. for liquids attacking borosilicate glass, Al2O3-ceramic, PFA, ETFE, FEP or PTFE 4. for Peroxide (due to catalytic reaction) 5. for carbon disulfide (CS2), due to risk of explosion!
Filling tubes for seripettor, available in lengths of 250 mm and 500 mm, made of polypropylene. With o-ring for sterile applications.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
1 of 1
Tato položka | |||
|---|---|---|---|
| description for Dispensette® III 0.5-10 mL | description PTFE, outer thread 3/4 in., GL 32 | description L 1 m, O.D. 6.9 mm | description L 10 m, O.D. 7.6 mm |
| material FEP | material PTFE | material FEP | material FEP |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| manufacturer/tradename BRAND 704209 | manufacturer/tradename BRAND 704281 | manufacturer/tradename BRAND 704269 | manufacturer/tradename BRAND 704267 |
Still not finding the right product?
Explore all of our products under BRAND® filling tube
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?