Skip to Content
Merck
All Photos(2)

Key Documents

WH0005690M2

Sigma-Aldrich

Monoclonal Anti-PSMB2 antibody produced in mouse

clone M1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-HC7I, Anti-MGC104215, Anti-MGC126885, Anti-proteasome (prosome, macropain) subunit, beta type, 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

M1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PSMB2(5690)

General description

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit. (provided by RefSeq)

Immunogen

PSMB2 (AAH00268, 1 a.a. ~ 201 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MEYLIGIQGPDYVLVASDRVAASNIVQMKDDHDKMFKMSEKILLLCVGEAGDTVQFAEYIQKNVQLYKMRNGYELSPTAAANFTRRNLADCLRSRTPYHVNLLLAGYDEHEGPALYYMDYLAALAKAPFAAHGYGAFLTLSILDRYYTPTISRERAVELLRKCLEELQKRFILNLPTFSVRIIDKNGIHDLDNISFPKQGS

Biochem/physiol Actions

Proteasome subunit β 2 (PSMB2) has a trypsin-like activity. It has a role in double-strand break repair and associates with homeobox A2 (HOXA2). Overexpression of the protein in human cells has been shown to reduce homologous recombination. It may trigger the loading of β subunit onto a rings in the proteasome.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The over-expression of the ?2 catalytic subunit of the proteasome decreases homologous recombination and impairs DNA double-strand break repair in human cells.
Collavoli A
Journal of Biomedicine and Biotechnology, 2011, 757960-757960 (2011)
The homeodomain transcription factor Hoxa2 interacts with and promotes the proteasomal degradation of the E3 ubiquitin protein ligase RCHY1.
Bergiers L
PLoS ONE, 8(11), e80387-e80387 (2013)
Differential intra-proteasome interactions involving standard and immunosubunits.
Jayarapu K and Griffin TA
Biochemical and Biophysical Research Communications, 358(3), 867-872 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service