Key Documents
CLS294775X50
Corning® microscope slides, plain
L × W 75 mm × 50 mm
Synonym(s):
microscope slides
About This Item
UNSPSC Code:
41121800
NACRES:
NB.02
material
soda-lime glass
sterility
non-sterile
packaging
case of 5 (pack 5)
manufacturer/tradename
Corning 2947-75X50
L × W
75 mm × 50 mm
thickness
0.90-1.10 mm
binding type
non-treated surface
General description
These 75 x 50 mm Corning microscope slides are made from water-white glass to maximize clarity and are not frosted. Slides are 0.9 to 1.10 mm thick.
Products are packaged in 1/2 gross boxes (approx. 72 slides each).
5 gross = 10 boxes (720 slides total)
Features and Benefits
  • Corning® microscope slides, plain are made of Type II - Soda-Lime Glass.
  • For general laboratory microscopy purpose.
  • Non-sterile.
  • Cannot be autoclaved and not intended for heating or cooling.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)