Skip to Content
Merck
All Photos(1)

Key Documents

18577

Sigma-Aldrich

Neryl monophosphate lithium salt

≥95.0% (TLC)

Synonym(s):

(Z)-3,7-Dimethyl-2,6-octadien-1-yl phosphate lithium salt, Neryl dihydrogen phosphate lithium salt, Neryl phosphate lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19O4P · xLi+
CAS Number:
Molecular Weight:
234.23 (free acid basis)
UNSPSC Code:
12352106
NACRES:
NA.25

Quality Level

Assay

≥95.0% (TLC)

storage temp.

−20°C

Related Categories

Application

Neryl phosphate, the cis isomer of geranyl phosphate, may be useful to study the kinetics of enzymes that use neryl or geranyl pyrophosphate as their substrates.

Biochem/physiol Actions

Monoterpene phosphate

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S M Stanley Fernandez et al.
Biochemistry, 39(50), 15316-15321 (2000-12-12)
Farnesyl diphosphate synthase (FPPase) catalyzes chain elongation of the C(5) substrate dimethylallyl diphosphate (DMAPP) to the C(15) product farnesyl diphosphate (FPP) by addition of two molecules of isopentenyl diphosphate (IPP). The synthesis of FPP proceeds in two steps, where the
M C Schulbach et al.
The Journal of biological chemistry, 276(15), 11624-11630 (2001-01-21)
We have recently shown that open reading frame Rv1086 of the Mycobacterium tuberculosis H37Rv genome sequence encodes a unique isoprenyl diphosphate synthase. The product of this enzyme, omega,E,Z-farnesyl diphosphate, is an intermediate for the synthesis of decaprenyl phosphate, which has
Hirekodathakallu V Thulasiram et al.
Journal of the American Chemical Society, 128(49), 15819-15823 (2006-12-07)
Farnesyl diphosphate (FPP) synthase catalyzes the consecutive head-to-tail condensations of isopentenyl diphosphate (IPP, C5) with dimethylallyl diphosphate (DMAPP, C5) and geranyl diphosphate (GPP, C10) to give (E,E)-FPP (C15). The enzyme belongs to a genetically distinct family of chain elongation enzymes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service