Skip to Content
Merck
All Photos(2)

Key Documents

72610

Sigma-Aldrich

3′-Nitroacetophenone

purum, ≥98.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4COCH3
CAS Number:
Molecular Weight:
165.15
Beilstein:
743002
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (GC)

form

crystals

bp

202 °C (lit.)

mp

76-78 °C (lit.)
77-80 °C

functional group

ketone
nitro

SMILES string

CC(=O)c1cccc(c1)[N+]([O-])=O

InChI

1S/C8H7NO3/c1-6(10)7-3-2-4-8(5-7)9(11)12/h2-5H,1H3

InChI key

ARKIFHPFTHVKDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

376.9 °F - closed cup

Flash Point(C)

191.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service