Skip to Content
Merck
All Photos(1)

Key Documents

1.00169

Supelco

di-Boron trioxide

99.9995 Suprapur®

Synonym(s):

di-Boron trioxide, Boric anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
B2O3
CAS Number:
Molecular Weight:
69.62
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-125-8
NACRES:
NB.21

grade

for inorganic trace analysis

Quality Level

Assay

≥99.0% (from ignited substance)
99.9995%

form

solid

potency

3150 mg/kg LD50, oral (Rat)

pH

4 (25 °C, 10 g/L in H2O)

mp

450 °C

solubility

36 g/L

density

2.46 g/cm3 at 20 °C

bulk density

900‑1100 kg/m3

anion traces

chloride (Cl-): ≤5 ppm
phosphate (PO43-): ≤5 ppm
silicate (as SiO2): ≤1 ppm
sulfate (SO42-): ≤5 ppm

cation traces

Ag: ≤0.020 ppm
Al: ≤0.050 ppm
As: ≤0.010 ppm
Au: ≤0.010 ppm
Ba: ≤0.020 ppm
Be: ≤0.010 ppm
Ca: ≤0.100 ppm
Cd: ≤0.010 ppm
Co: ≤0.010 ppm
Cr: ≤0.020 ppm
Cu: ≤0.020 ppm
Fe: ≤0.050 ppm
In: ≤0.010 ppm
K: ≤0.20 ppm
Li: ≤0.010 ppm
Mg: ≤0.050 ppm
Mn: ≤0.010 ppm
Mo: ≤0.020 ppm
Na: ≤0.20 ppm
Ni: ≤0.020 ppm
Pb: ≤0.020 ppm
Sr: ≤0.020 ppm
Tl: ≤0.010 ppm
V: ≤0.020 ppm
Zn: ≤0.050 ppm

storage temp.

2-30°C

SMILES string

B(=O)OB=O

InChI

1S/B2O3/c3-1-5-2-4

InChI key

JKWMSGQKBLHBQQ-UHFFFAOYSA-N

Related Categories

Application


  • Raman and Infrared Spectroscopy of Barium-Gallo Germanate Glasses Containing B(2)O(3)/TiO(2).: This study utilizes Raman and infrared spectroscopy to analyze the structural properties of barium-gallo germanate glasses containing di-Boron trioxide (B2O3). These insights are crucial for developing new materials with tailored properties for various industrial applications (Kowalska et al., 2023).

  • Transition Metals (Cr(3+)) and Lanthanides (Eu(3+)) in Inorganic Glasses with Extremely Different Glass-Formers B(2)O(3) and GeO(2).: This research investigates the incorporation of transition metals and lanthanides in inorganic glasses formed with di-Boron trioxide, enhancing their optical properties and potential applications in photonics and other advanced materials (Kowalska et al., 2021).

  • Synthesis and characterization of Ag(2)O/B(2)O(3)/TiO(2) ternary nanocomposites for photocatalytic mineralization of local dyeing wastewater under artificial and natural sunlight irradiation.: This research presents a novel method for synthesizing nanocomposites involving di-Boron trioxide (B2O3), with significant implications for environmental applications, particularly in wastewater treatment (Tijani et al., 2019).

Analysis Note

Purity (metallic): ≥ 99.9995 %
Assay (from ignited substance): ≥ 99.0 %
Chloride (Cl): ≤ 5 ppm
Phosphate (PO): ≤ 5 ppm
Silicate (as SiO): ≤ 1 ppm
Sulfate (SO): ≤ 5 ppm
Ag (Silver): ≤ 0.020 ppm
Al (Aluminium): ≤ 0.050 ppm
As (Arsenic): ≤ 0.010 ppm
Au (Gold): ≤ 0.010 ppm
Ba (Barium): ≤ 0.020 ppm
Be (Beryllium): ≤ 0.010 ppm
Ca (Calcium): ≤ 0.100 ppm
Cd (Cadmium): ≤ 0.010 ppm
Co (Cobalt): ≤ 0.010 ppm
Cr (Chromium): ≤ 0.020 ppm
Cu (Copper): ≤ 0.020 ppm
Fe (Iron): ≤ 0.050 ppm
In (Indium): ≤ 0.010 ppm
K (Potassium): ≤ 0.20 ppm
Li (Lithium): ≤ 0.010 ppm
Mg (Magnesium): ≤ 0.050 ppm
Mn (Manganese): ≤ 0.010 ppm
Mo (Molybdenum): ≤ 0.020 ppm
Na (Sodium): ≤ 0.20 ppm
Ni (Nickel): ≤ 0.020 ppm
Pb (Lead): ≤ 0.020 ppm
Sr (Strontium): ≤ 0.020 ppm
Tl (Thallium): ≤ 0.010 ppm
V (Vanadium): ≤ 0.020 ppm
Zn (Zinc): ≤ 0.050 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service