Z755524
Mini BlotBoy™ 3D platform rocker
AC/DC input 230 V AC, European 2-pin plug, CE compliant
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
tilt angle: +/- 8°
feature
CE compliant
manufacturer/tradename
Benchmark Scientific B3D1008-230-EU plug
parameter
12 rpm speed (10 rpm for 230V)
4-65 °C temp. range
4.5 lb load
W × D × H
10.5 in. (27 cm) × 7.5 in. (19 cm) × 8 in. (20 cm)
platform L × W
10.5 in. (27 cm) × 7.5 in. (19 cm)
Looking for similar products? Visit Product Comparison Guide
General description
The Mini BlotBoy platform rockers provide the perfect 3-dimensional motion for all blot related applications. The tilt angle and speed have been optimally set for gentle, but thorough mixing in gel trays, boxes and other flat vessels.
Mini BlotBoys are supplied with autoclavable flat mats and are ideal for blot washes and gel staining/destaining. Designed to conserve bench space, the Mini BlotBoy is supplied with a 10.5 x 7.5 inch platform. The Mini BlotBoy can accept stackable platforms, handles loads up to 2kg, and has maintenance free brushless motors; safe for use in cold rooms and incubators.
Mini BlotBoys are supplied with autoclavable flat mats and are ideal for blot washes and gel staining/destaining. Designed to conserve bench space, the Mini BlotBoy is supplied with a 10.5 x 7.5 inch platform. The Mini BlotBoy can accept stackable platforms, handles loads up to 2kg, and has maintenance free brushless motors; safe for use in cold rooms and incubators.
Legal Information
BlotBoy is a trademark of Benchmark Scientific, Inc.
Mini BlotBoy is a trademark of Benchmark Scientific, Inc.
accessory
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service