Skip to Content
Merck
All Photos(2)

Key Documents

Z680540

Duran® pressure equalization set

for 4-port cap, GL 45 (0.2 μm incl. membrane filter)

Synonym(s):

1137801

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

description

with 0.2 μm membrane filter

manufacturer/tradename

DURAN® 1137801

Looking for similar products? Visit Product Comparison Guide

General description

Assembled set for pressure equalization, has screw cap GL 45 with three or four ports that have GL 14 thread.

Features and Benefits

• Suitable for two different HPLC tubing outer diameters 1.6 or 3.2 mm• Unused ports can be sealed with silicone closure / blanking seals• Optional sterile pressure equalisation set (25 mm syringe filter 0.2 µm)• Caps block solvent vapours VOCs, and keep your mobile phase clean. They improve lab health and safety, and the quality of your analysis

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service