Key Documents
Z680400
Duran® GL 45 connection system caps and accessories
GL 45 blue PP screw cap with 3 x GL 14 ports
Synonym(s):
1129751, Connection System Accessories, Duran Connection Caps, GL 45 Connection System
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
blue-gray polypropylene
manufacturer/tradename
DURAN® 1129751
parameter
-40-140 °C temp. limit
General description
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available: 1.6 mm, 3.0 mm, 3.2 mm and 6.0 mm that can be connected.
Sterile pressure equalization is possible through use of membrane filter.
Features and Benefits
• Manufactured from Polypropylene.• Rigid or semi-rigid tubing of different diameters can be accepted by using a blue GL 14 open topped cap with one of the inserts.• Available with 2, or 3 threaded outlets (ports).• Four different tubing diameters (1.6 mm, 3.0 mm, 3.2 mm or 6.0 mm tubing outer diameter) can be used by changing the GL 14 inserts.• By applying positive or negative pressure liquid can be pumped out of or into the bottle via the screw cap.• Fully compatible with all the GL 45 threads and therefore highly versatile.
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)