Key Documents
Z304816
Quickfit® powder funnels
joint: ST/NS 34/35
Synonym(s):
CF5/5
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
Joint
joint: ST/NS34/35
manufacturer/tradename
Quickfit CF 5/5
availability
not available in USA and Canada
General description
For pouring powders or liquids into vessels with ground-glass joints
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)