Key Documents
SYNF110350
Synthware Dewar flask, tall form with metal housing
flask capacity 350 mL, tall form with metal housing
Synonym(s):
Synthware F110350
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
material
aluminum flask
Joint
Joints neck (70)
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F110350
I.D. × depth
7 cm × 12.5 cm
flask capacity
350 mL
General description
Dewar flask with weighted metal base for stability and with a full-length aluminum housing which extends to the top rim for added protection.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)