Skip to Content
Merck
All Photos(1)

Key Documents

BR148025

BRAND® PP powder funnel short wide stem

top diam. 80 mm, bottom diam. 15 mm

Synonym(s):

Funnels, Powder funnels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.43

packaging

pack of 20 ea

manufacturer/tradename

BRAND 148025

bottom diam.

15 mm

top diam.

80 mm

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • BRAND® powder funnel is ideal for transfering powders.
  • Made from break-resistant polypropylene, these general lab use funnels include a short wide stem that facilitates optimal filtration.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service