BR137638
BRAND® magnetic stirring bar, PTFE, crosshead
crosshead, H × D 15 mm × 38 mm
Synonym(s):
Magnetic Stirring Bars, PTFE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PTFE-coated
crosshead
manufacturer/tradename
BRAND 137638
H × D
15 mm × 38 mm
L
8 mm
diam.
15 mm
Looking for similar products? Visit Product Comparison Guide
General description
Steady spinning position. Excellent stirring action.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service