Z284238
Spinstar® magnetic stirring bar
fits beaker size, 400 mL
Synonym(s):
Stir Bar, Stirbar, Stirring Bar
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PTFE-coated (Star)
PTFE-coated
cross
feature
fits beaker size 400 mL
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37146-0400
bar I.D.
70 mm
diam.
70 mm
Looking for similar products? Visit Product Comparison Guide
General description
Star-shaped bard create a deep vortex at slow speeds.
Legal Information
Spinstar is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service