Key Documents
BAF188700000
SP Bel-Art® Radioimmunoassay Tube Rack
to hold, 216 x 10-13mm Tubes
Synonym(s):
polypropylene rack, tube holder
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
feature
to hold 216 x 10-13mm Tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18870-0000
H × L × width
3 in. (76 mm) × 13.5 in. (342 mm) × 9.0625 in. (230 mm)
platform H
2.25 in. (57 mm)
General description
These polypropylene racks provide a practical method of pipetting and storing a large number of samples.
Features and Benefits
  • Easy tracking of last tube filled by using alternate rows and rotating the tubes
  • Useful in clinical labs for storage of samples
  • Steam autoclavable at 121ºC (250ºF)
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)