Key Documents
BAF185120000
SP Bel-Art® Scintillation Vial Rack
to hold, holds 24 x 25-30mm Vials
Synonym(s):
autoclavable polypropylene
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
feature
to hold holds 24 x 25-30mm Vials
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18512-0000
H × L × W
3.75 in. (95 mm) × 11.75 in. (298 mm) × 5 in. (127 mm)
General description
This rugged and stable rack holds 24 scintillation vials (30mm diameter) for convenient storage, handling, and sample preparation.
Features and Benefits
  • This rugged and stable rack holds 24 scintillation vials (30mm diameter) for convenient storage, handling, and sample preparation.
  • Corrosion-resistant, autoclavable polypropylene
  • Shelf height: 4.4cm (1³/₄")
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)