Skip to Content
Merck
All Photos(1)

Key Documents

932930

Sigma-Aldrich

Tweezer

High Precision, Style 0

Synonym(s):

Gripper, Tweezer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

General description

EMS techware

Application

For the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips. Made from antimagnetic/anti-acid steel with smooth surface. Tips are PEEK/CF30. Ideal for gently holding wafers.

Tip Width: 12 mm. Depth: 2.0 mm.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service