Skip to Content
Merck
All Photos(1)

Key Documents

932787

Sigma-Aldrich

Tweezers

Style 5TTH

Synonym(s):

Tweezer, Grippers, Tweezers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

General description

EMS techware

Application

High precision and ultrafine tweezers. Typical applications include tweezers for the electronic industry, watch-makers, jewelers and laboratory applications in moderately aggressive chemical environments. Anti-capillary, extra fine tips with locking ring. Useful for handling EM grids. Length: 110mm.
The tweezers are the Swiss highest quality. Antimagnetic/anti-acid SA low carbon austenitic steel tweezers are corrosion resistant. Anti-glare satin finish. Useful for handling EM grids.
Material: SA Steel. OAL: 110 mm. Tips: Anti-capillary, extra fine tips with locking ring.
To improve your productivity and allow you to view more than one sample at a time

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service