Skip to Content
Merck
All Photos(1)

Key Documents

900036

Sigma-Aldrich

Sodium taurocholate-2,2,4,4-d4

≥98 atom % D, ≥98% (CP)

Synonym(s):

Taurocholic-2,2,4,4-d4 acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26D4H40NaNO7S
Molecular Weight:
541.71
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+4

storage temp.

−20°C

SMILES string

[2H]C1([2H])[C@H](O)C([2H])([2H])C(C[C@@H](O)C2C3C[C@H](O)[C@@]4(C)C2CC[C@H]4[C@@H](CCC(NCCS(=O)([O-])=O)=O)C)[C@]3(C)C1.[Na+]

InChI

1S/C26H45NO7S.Na/c1-15(4-7-23(31)27-10-11-35(32,33)34)18-5-6-19-24-20(14-22(30)26(18,19)3)25(2)9-8-17(28)12-16(25)13-21(24)29;/h15-22,24,28-30H,4-14H2,1-3H3,(H,27,31)(H,32,33,34);/q;+1/p-1/t15-,16?,17+,18?,19?,20?,21-,22+,24?,25+,26-;/m1./s1/i8D2,12D2;

InChI key

JAJWGJBVLPIOOH-FAHHLWHHSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service