Skip to Content
Merck
All Photos(1)

Key Documents

614130

Sigma-Aldrich

Deoxycholic acid-2,2,4,4-d4

≥98 atom % D, ≥98% (CP)

Synonym(s):

3a,12a-Dihydroxy-5ß-cholanic acid-2,2,4,4-d4, 7-Deoxycholic acid,-2,2,4,4-d4, Desoxycholic acid-2,2,4,4-d4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24D4H36O4
CAS Number:
Molecular Weight:
396.60
MDL number:
UNSPSC Code:
51113400
PubChem Substance ID:

isotopic purity

≥98 atom % D

Assay

≥98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mp

171-174 °C

mass shift

M+4

SMILES string

[H][C@@]12[C@]([C@](CC([2H])([2H])[C@@H](O)C3([2H])[2H])(C)[C@]3([H])CC2)([H])C[C@H](O)[C@@]4(C)[C@@]1([H])CC[C@]4([H])[C@]([H])(C)CCC(O)=O

InChI

1S/C24H40O4/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3/h14-21,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-/m1/s1/i10D2,12D2

InChI key

KXGVEGMKQFWNSR-FCSCGBJGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tannaz Ghaffarzadegan et al.
Scientific reports, 9(1), 3800-3800 (2019-03-09)
Bile acids (BAs) are known to be involved in cholesterol metabolism but interactions between the diet, BA profiles, gut microbiota and lipid metabolism have not been extensively explored. In the present study, primary and secondary BAs including their glycine and
P Franco et al.
Journal of chromatography. A, 1585, 70-81 (2018-11-30)
Bile acids (BAs) are endogenous steroids involved in the transport of lipids in bile, acting also as molecular signaling hormones. Primary BAs synthesized in the liver undergo several metabolic pathways in the intestine by gut microbiota to produce secondary BAs.
Limin Zhang et al.
Environmental health perspectives, 123(7), 679-688 (2015-03-15)
Alteration of the gut microbiota through diet and environmental contaminants may disturb physiological homeostasis, leading to various diseases including obesity and type 2 diabetes. Because most exposure to environmentally persistent organic pollutants (POPs) occurs through the diet, the host gastrointestinal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service