Skip to Content
Merck
All Photos(1)

Key Documents

706698

Sigma-Aldrich

Allyl-13C3-amine-15N

99 atom % 13C, 98 atom % 15N, 98% (CP)

Synonym(s):

2-Propen-13C3-1-ylamine-15N, 3-Amino-15N-1-propene-13C3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H213C=13CH13CH215NH2
CAS Number:
Molecular Weight:
61.07
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % 15N

Assay

98% (CP)

form

liquid

bp

53 °C

density

0.813 g/mL at 25 °C

mass shift

M+4

SMILES string

[15NH2][13CH2][13CH]=[13CH2]

InChI

1S/C3H7N/c1-2-3-4/h2H,1,3-4H2/i1+1,2+1,3+1,4+1

InChI key

VVJKKWFAADXIJK-JCDJMFQYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service